Imprimer
Adrenergic ligand library, Screen-Well®
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BML28110100 BML28110500
ENZOBML28110100EA 860 EUR
ENZOBML28110100 ENZOBML28110500
Adrenergic ligand library, Screen-Well®
Suppléments et additifs pour culture cellulaire
The adrenergic ligand library contains 84 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein-coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The adrenergic ligand library is 1 plate of the larger neurotransmitter library which contains over 650 CNS receptor ligands.