Imprimer
Neurotransmitter library (10 plate set), Screen-Well®
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BML28100100 BML28100500
ENZOBML28100100EA 10020 EUR
ENZOBML28100100 ENZOBML28100500
Neurotransmitter library (10 plate set), Screen-Well®
Suppléments et additifs pour culture cellulaire
The Screen-Well® neurotransmitter library contains 681 CNS receptor ligands in a 96-well format. Ideal for screening or identifying recombinant orphan G protein-coupled receptors, target validation, secondary screening, validating new assays, and for routine pharmacological applications.
Includes 13 classes of receptor ligands in 10 deep-well plates (Adrenergis; Histaminergics, Melatonin ligands; Dopaminergics; Ionotropic; Glutamatergics; Serotonergics; Metabotropic; Glutamatergics; Opioids, Sigma ligands; GABAergics; Cholinergics; Purinergics; Adenosines). Each ligand is supplied in solution in a biocompatible solvent at a standard concentration of 10 mM (Opioid peptides are supplied at a concentrations of 1 mM). Plates are available as a complete set, or individually. Supporting data are included.