Imprimer
Ac-Leu-Leu-Nle-CHO
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BMLP1200005 BMLP1200025
ENZOBMLP1200005EA 116 EUR
ENZOBMLP1200005 ENZOBMLP1200025
Ac-Leu-Leu-Nle-CHO
Protéines et Peptides
Cysteine protease inhibitor
Cell-permeable, peptide aldehyde inhibitor of calpain I (Ki=190 nM), calpain II (Ki=150 nM), cathepsin L (Ki=0,5 nM) and other neutral cysteine proteases. Inhibits cell cycle progression at G1/S and metaphase/anaphase in CHO cells by inhibiting cyclin B degradation. Also stimulates HMG-CoA synthase transcription by inhibiting degradation of active SREBP-1 (sterol regulatory element-binding protein 1). Protects against neuronal damage caused by hypoxia and ischemia. Inhibits apoptosis in thymocytes and metamyelocytes. Also prevents nitric oxide production by activated macrophages by interfering with the transcription of inducible nitric oxide synthase (iNOS; NOS II). Inhibits proteolytic degradation of IκBα and IκBß in RAW macrophages induced with LPS. It also prolong association of MHC class I molecules with the transporters associated with antigen processing.