Imprimer
Phosphatase inhibitor library, Screen-Well®
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BML28340100 BML28340500
ENZOBML28340100EA 1580 EUR
ENZOBML28340100 ENZOBML28340500
Phosphatase inhibitor library, Screen-Well®
Suppléments et additifs pour culture cellulaire
The Screen-Well™ Phosphatase Inhibitor Library contains 33 known phosphatase inhibitors of well-defined activity. Inhibitors are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates at either 100 µl or 500 µl per well. The library is ideal for chemical genomics, assay development, and as a reference set for secondary screening. Includes inhibitors of these important phosphatases: Calcineurin(PP2B), JSP-1, PRL-1, CD45, PP1, PRL-3, CDC25, PP2A, PTEN, and many more.