Imprimer
Z-VRPR-FMK
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ALX260166M001 ALX260166MC05
ENZOALX260166M001EA 753 EUR
ENZOALX260166M001 ENZOALX260166MC05
Z-VRPR-FMK
Protéines et Peptides
MALT1 inhibitor
The proteolytic activity of the paracaspase MALT1 is key to NF-κB and T cell activation. Z-VRPR-FMK is a selective MALT1 inhibitor of such proteolytic activity.