Imprimer
Human recombinant FGF-basic 147
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ALX2018220010 ALX2018220050
ENZOALX2018220010EA 204 EUR
ENZOALX2018220010 ENZOALX2018220050
Human recombinant FGF-basic 147
Protéines et Peptides
Produced in E. coli. Non-glycosylated protein, containing 147 amino acids.
  • Carrier-free