Imprimer
Salmon Calcitonin
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BMLP4200001 BMLP4200005
ENZOBMLP4200001EA 139 EUR
ENZOBMLP4200001 ENZOBMLP4200005
Salmon Calcitonin
Protéines et Peptides
Reduces blood calcium
Calcium-regulating peptide hormone released from the thyroid. Lowers calcium concentration in the blood and decreases bone resorption by osteoclasts. Used for therapeutic applications in osteoporosis. Salmon calcitonin peptide sequence differs from mammalian but is more potent.