Imprimer
Ac-LEVD-AFC
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ALX260084M001 ALX260084M005
ENZOALX260084M001EA 93.5 EUR
ENZOALX260084M001 ENZOALX260084M005
Ac-LEVD-AFC
Protéines et Peptides
Caspase-4 substrate
Fluorogenic substrate for caspase-4. Similar to the fluorogenic substrate Ac-LEVD-AMC (Prod. No. ALX-260-083); the AFC fluorophore is more sensitive.