Imprimer
Ac-YVAD-pNA
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ALX260026M001 ALX260026M005
ENZOALX260026M001EA 68.2 EUR
ENZOALX260026M001 ENZOALX260026M005
Ac-YVAD-pNA
Protéines et Peptides
Caspase-1 substrate
Sequence is a modification of pro-interleukin-1beta Asp-116 cleavage site for caspase-1. Similar to Ac-YVAD-AMC but cleavage is monitored colorimetrically at 405 nm.