Imprimer
PBS (endotoxin-free), Enzo Life Sciences
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ALX505007L010 ALX505007LD15
ENZOALX505007L010EA 165 EUR
ENZOALX505007L010 ENZOALX505007LD15
PBS (endotoxin-free), Enzo Life Sciences
Tampons Tampons de culture de tissus
Sterile and endotoxin-free PBS for use with TLRgrade™ or endotoxin-free grade reagents.
Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal.