Imprimer

Anti-HSP90B1 Rat Monoclonal Antibody [clone: 9G10]

Fournisseur: ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ADISPA850488D ADISPA850488F
ENZOADISPA850488DEA 321 EUR
ENZOADISPA850488D ENZOADISPA850488F
Anti-HSP90B1 Rat Monoclonal Antibody [clone: 9G10]
Anticorps
Glucose-regulated protein 94 (Grp94, gp96), an abundant resident endoplasmic reticulum (ER) lumenal stress protein, belongs to the Hsp90 family of molecular chaperones along with cytosolic Hsp90. Grp94 and such other resident soluble proteins of the ER as the Ca 2+ binding protein subfamily (CaBP, CaBPI, CaBP2 and calreticulin) possess the C-terminal tetrapeptide Lys-Asp-Glu-Leu (KDEL), a sorting signal considered responsible for the retention of these proteins in the pre-Golgi compartments. Stress conditions such as glucose starvation and heat shock which promote protein misfolding or unfolding increase Grp94 expression. In addition to a homeostatic role in protein folding and assembly, Grp94 can function in the intracellular trafficking of peptides from the extracellular space to the MHC class I antigen processing pathway of antigen presentation cells. Grp94 and Hsp90 share high sequence identity and apparently identical adenosine nucleotide dependent modes of regulation, although previous data suggests that Hsp90 and Grp94 may differ in their nucleotide binding properties. The N-terminal domain of eukaryotic Hsp90 proteins contains a conserved adenosine nucleotide binding pocket which also serves as the binding site for the Hsp90 inhibitors geldanamycin and radicicol. However, the molecular basis for adenosine nucleotide-dependent regulation of Grp94 remains unclear. Data supports a ligand dependent regulation of Grp94 function, and suggests a model whereby Grp94 function is regulated through a ligand -dependent conversion of Grp94 from an inactive to an active conformation.

Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: 9G10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Pig, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR