Imprimer

Anti-VAMP2 Rabbit Polyclonal Antibody

Fournisseur: ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ADIVASSV006D ADIVASSV006F
ENZOADIVASSV006DEA 235 EUR
ENZOADIVASSV006D ENZOADIVASSV006F
Anti-VAMP2 Rabbit Polyclonal Antibody
Anticorps
Vesicle-associated membrane protein (VAMP), also known as synaptobrevin, is an 18 kDa integral membrane protein localized to the cytoplasmic surface of synaptic vesicles. Two VAMP homologs, VAMP1 and VAMP2, are differentially expressed in the nervous system. VAMP2 is found in neurons associated with autonomic, sensory and integrative functions, and in non-neuronal Langerhans islets and glomerular cells. VAMP is a component of a 20S complex implicated in vesicle docking and fusion, the formation of which involves the interaction of VAMP with syntaxin through an ATP-dependent reaction modulated by synaptotagmin, SNAP-25, alpha-SNAP, and NSF.

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, guinea pig, and rabbit VAMP2. Detects a band of ~18kDa by Western blot.

Type: Primary
Antigen: VAMP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR