Imprimer

Anti-Membrin Mouse Monoclonal Antibody

Fournisseur: ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ADIVAMPT046D ADIVAMPT046F
ENZOADIVAMPT046DEA 282 EUR
ENZOADIVAMPT046D ENZOADIVAMPT046F
Anti-Membrin Mouse Monoclonal Antibody
Anticorps
Membrin, also known as GS27 (Golgi SNARE of 27K), is an integral membrane protein present on the surface of the Golgi apparatus. GS27 is a member of a protein complex that consists of cis-Golgi vesicle receptor syntaxin 5 and GS28, v-SNARE rbet1 and Rsec22, and Rsly1. These complexes are postulated to direct the fusion of ER-derived vesicles with vesicular tubular cluster (VTC), and the fusion of VTCs to form cis-Golgi compartment.

  • For Immunocytochemistry, Western Blot

Recognizes human, rat, dog, chicken, hamster, monkey, pig and rabbit Membrin. Detects a band of ~27kDa by Western blot.

Type: Primary
Antigen: MEMBRIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR