Imprimer

Anti-MT Mouse Monoclonal Antibody

Fournisseur: ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ADISPA550D ADISPA550F
ENZOADISPA550DEA 290 EUR
ENZOADISPA550D ENZOADISPA550F
Anti-MT Mouse Monoclonal Antibody
Anticorps
Metallothioneins (MTs) are a class of ubiquitous, low molecular weight, cysteine-rich, metal binding proteins that function in the metabolism of zinc and copper. MT protects against exposure to toxic metals such as mercury and cadmium as well as a variety of other stress conditions. There are four mammalian MT isoforms: MT-I, -II, -III, -IV, each of which is encoded by individual genes. MT expression is induced by heavy metal cations, starvation, elevated temperature, lipopolysaccharide, inflammatory cytokines, free radicals, and ionizing radiation.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human, mouse, rat, fish, horse and rabbit metallothionein. Detects a band of ~15kDa by Western blot.

Type: Primary
Antigen: MT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR