Imprimer

Anti-Cu/Zn SOD Rabbit Polyclonal Antibody

Fournisseur: ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ADISOD100D ADISOD100F ADISOD100J
ENZOADISOD100DEA 267 EUR
ENZOADISOD100D ENZOADISOD100F ENZOADISOD100J
Anti-Cu/Zn SOD Rabbit Polyclonal Antibody
Anticorps
SOD (Superoxide dismutase) is responsible for the elimination of cytotoxic active oxygen by catalyzing the dismutation of the superoxide radical to oxygen and hydrogen peroxide. There are three SOD isoenzymes in mammalian cells, they are: EC SOD (extracellular SOD), Cu/Zn SOD (copper and zinc-containing SOD) and Mn SOD (manganese-containing SOD).

  • For Immunofluorescence, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, and clam Cu/Zn SOD. Detects a band of ~23kDa (human) or ~19kDa (mouse and rat) by Western blot.

Type: Primary
Antigen: Cu/Zn SOD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR