Imprimer

Anti-RAS Rabbit Polyclonal Antibody

Fournisseur: ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ADIKAPGP001D ADIKAPGP001F
ENZOADIKAPGP001DEA 225 EUR
ENZOADIKAPGP001D ENZOADIKAPGP001F
Anti-RAS Rabbit Polyclonal Antibody
Anticorps
Monomeric G-protein Ras functions as a molecular switch linking receptor and nonreceptor tyrosine kinase activation to a number of downstream signaling pathways. Oncogenic mutations, which occur in approximately 30% of all cases of human cancer, lead to the stimulation of proliferation and the inhibition of apoptosis.

  • For Western Blot

Recognizes human, mouse, rat and bovine Ras. Detects a band of 21-23kDa by Western blot.

Type: Primary
Antigen: RAS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR