Imprimer
Bags, bin box, B-Safe
Fournisseur: Alpha Packaging
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
526089 526090 526091
129-0403EA 164 EUR
129-0403 129-0404 129-0405
Bags, bin box, B-Safe
Sachets Sacs à déchets
These bags are made from PE, 125 µm (500 gauge), clear.
  • Heat sealed for leak protection
  • Each bag is box-shaped and has a double-string neck pull
  • No printing
Informations de livraison: Supplied in benchtop dispenser boxes.