Imprimer
Bottles, Narrow Neck, Cylindrical, Round, HDPE, Rixius
Fournisseur: RIXIUS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3010010002855W 3010001001810 3-0400-0250-28 3-010010002515 30100025018180 30100010028WC 41001200013WC
RIXI3010010002855WEA 1740 EUR
RIXI3010010002855W RIXI3010001001810 RIXI3-0400-0250-28 RIXI3-010010002515 RIXI30100025018180 RIXI30100010028WC RIXI41001200013WC
Bottles, Narrow Neck, Cylindrical, Round, HDPE, Rixius
Flacons Flacons en plastique
Made of HDPE.
  • With or without closure
  • Good chemical resistance
  • Good temperature resistance
Cylindrical round bottles.
Informations de commande: Closures must be ordered separately.
Attention:
*The product(s) on this page are outside the scope of the EU Directive 2019/904.