Imprimer
Reducing pieces for NW flanges
Fournisseur: EDWARDS HIGH VACUUM METTLER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C10516449 C10514446 C10514447 C10516448
EDWAC10516449EA 209 EUR
EDWAC10516449 181-0368 EDWAC10514447 EDWAC10516448
Reducing pieces for NW flanges
Raccords et connecteurs
Reducing pieces made of stainless steel.
Certifications: AISI 316L DIN 1.4404