Imprimer
Biological indicators, spore threads
Fournisseur: CROSSTEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
THS-06 THS-06P
NAMSTHS-06EA 792 EUR
NAMSTHS-06 NAMSTHS-06P
Biological indicators, spore threads
Indicateurs de stérilisation
Spore threads are prepared from Geobacillus stearothermophilus. These are designed to be placed directly into a device and utilised to monitor steam sterilisation process efficacy.
Ideal for use in tubing, lumens and other devices/instruments where larger BIs and carriers cannot be utilised.