Imprimer
Pump tubing, Tygon® MHLL 2765-175
Fournisseur: Saint Gobain Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
07MHLL-18 07MHLL-19
VERN07MHLL-18EA 287 EUR
VERN07MHLL-18 VERN07MHLL-19
Pump tubing, Tygon® MHLL 2765-175
Tuyaux Transfer Tubing
Longest flex life of any organic solvent resistant tubing.
  • Plasticizer free
  • Virtually no extractables
Especially designed for MEK and other aggressive solvents.
Temperature resistance: −70 to +74 °C
Hardness (Shore durometer A): 64
Certifications: Complies with US Pharmacopoea, Class VI, US Pharmacopoea, Klasse VI, FDA 21CFR 177.2600 criterias.