Imprimer
Cell culture plates, OneWell Plate™, CELLSTAR®
Fournisseur: Greiner Bio-One
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
670180_32 670190_32
GREI670180_32EA 354 EUR
GREI670180_32 GREI670190_32
Cell culture plates, OneWell Plate™, CELLSTAR®
Microplaques Microplaques pour culture cellulaire
The CELLSTAR® OneWell Plate™ is designed for use in applications involving the cultivation of large quantities of cells. Made from PS.
  • Free from RNase, DNase, and human DNA
  • Non pyrogenic and non cytotoxic
The plate is suitable for use with a wide range of automated cell culture and liquid handling systems. The rectangular shape of the CELLSTAR® OneWell Plate™ allows for an efficient usage of incubator space. Notches on the left side of the plate and the lid ensure a secured lid position.
L×W×H: 127,8×85,5×14,4 mm
Certifications: Complies with ANSI standards.
Informations de livraison: Supplied with a PS lid.