Imprimer
Chromatography tubing, PTFE
Fournisseur: VICI JOUR RESEARCH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
JR-T-4011C-M3 JR-T-4036C-M3 JR-T-4183C-M3 JR-T-4011C-M10 JR-T-4011C-M25 JR-T-4183C-M10 JR-T-4036C-M25 JR-T-4036C-M10 JR-T-4183C-M25
JOURJR-T-4011C-M3EA 45.7 EUR
JOURJR-T-4011C-M3 JOURJR-T-4036C-M3 JOURJR-T-4183C-M3 JOURJR-T-4011C-M10 JOURJR-T-4011C-M25 JOURJR-T-4183C-M10 JOURJR-T-4036C-M25 JOURJR-T-4036C-M10 JOURJR-T-4183C-M25
Chromatography tubing, PTFE
Tuyaux Tubulures à chromatographie
These PTFE tubings are biocompatible, chemically inert and suitable for lower or mild pressure applications.
  • Tolerance: ±0,05 mm