Imprimer
Handheld digital microscopes, Dino-Lite High Speed (USB 3.0)
Fournisseur: Dino-Lite / IDCP B.V.
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AM73115MZT AM73115MZTL AM73915MZT AM73915MZTL AM73515MZT AM73515MZTL AM73115MTF
IDCPAM73115MZTEA 1240 EUR
IDCPAM73115MZT IDCPAM73115MZTL IDCPAM73915MZT IDCPAM73915MZTL IDCPAM73515MZT IDCPAM73515MZTL IDCPAM73115MTF
Handheld digital microscopes, Dino-Lite High Speed (USB 3.0)
Microscopes
Dino-Lite models with USB 3.0 offer an image transfer speed of up to 45 fps at a 1280×960 resolution. USB 3.0 adds a new transfer mode called "SuperSpeed" (SS) capable of transferring data up to 5 Gbits/s (625 MB/s). SuperSpeed allows users to take advantage of the full power of the Dino-Lite Edge Series optics including improved colour accuracy and image quality.
  • Super speed USB 3.0 connection
  • Metal housing: Microscopes are ESD-safe
  • Edge series: High-quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
  • Integrated polariser greatly reduces glare on reflective surfaces (included with all models except AM73115MTF)
  • Model AM73115MTF: Extra Long Working Distance (ELWD) of up to 48 cm
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Models with Flexible LED control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant.
Informations de livraison: Supplied with carrying pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)), USB cable, user manual, quick guide, calibration target, and front cover box.
Attention: Please note: Dino-Lite USB 3.0 models use a custom USB cable that is created specifically for Dino-Lite.