Imprimer
Reaction flask, with four necks
Fournisseur: QVF LABORTECHNIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
KOK01032CV06 KOK01032CV07 KOK01033AV07 KOK01033AV08
QVFLKOK01032CV06EA 0 EUR
QVFLKOK01032CV06 QVFLKOK01032CV07 QVFLKOK01033AV07 QVFLKOK01033AV08
Reaction flask, with four necks
Ballons Flacons à fond rond
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These reaction flasks have three necks with 7° distance from the centre neck. Manufactured according to DIN 12 392.