Imprimer
Microplate sieves
Fournisseur: ENDECOTTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
100SMP.005 100SMP.020 100SMP.050 003SMP.005 008SMP.005 200SMP.005 200SMP.010 200SMP.015
ENDE100SMP.005EA 2850 EUR
ENDE100SMP.005 ENDE100SMP.020 ENDE100SMP.050 ENDE003SMP.005 ENDE008SMP.005 ENDE200SMP.005 ENDE200SMP.010 ENDE200SMP.015
Microplate sieves
Tamis Tamis analytiques
These microplate sieves are made from electro-formed nickel plate in stainless steel frames of 100 mm or 200 mm diameter.
  • Precision frame ensures consistent nestability
  • Natural fillet for free flowing of sample
  • Totally sealed with no crevice to lose material
  • Evenly tensioned mesh which ensures accurate analysis
Features unique self clearing apertures for the sizes from 75 to 5 microns. It is recommended that microplate sieves are used in conjunction with a liquid medium to assist the passage of extremely fine particles through the apertures. InEn savoir plus