Imprimer
Brushes for tubes and barrels
Fournisseur: Socorex Isba
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.170.320 1.170.305 1.170.300
SOCO1.170.320EA 32.6 EUR
SOCO1.170.320 SOCO1.170.305 SOCO1.170.300
Brushes for tubes and barrels
Brosses
Cylinder and tube brushes with tufted end and plastic handle for glass barrel and tube cleaning.
Informations de livraison: The set includes: Two brushes for 0,3/0,5/1 ml barrels, two brushes fpr 1/2/5 ml barrels and two brushes for 5/10/20 ml barrels.