Imprimer
Bottles, wide neck, HDPE, with sealing cap
Fournisseur: LAMAPLAST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
343 344 346
LAMA343EA 2.9 EUR
LAMA343 LAMA344 LAMA346
Bottles, wide neck, HDPE, with sealing cap
Flacons Flacons en plastique
These HDPE bottles are suitable for the strorage of dense fluids, granulates and powders.
  • Complete emptying is guaranteed by the inclination of the shoulders
  • Screw cap with opening-off seal
  • For food use