Imprimer
Kjeldahl weighing boat
Fournisseur: VELP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CM0486000 CM0486001
VELPCM0486000EA 210 EUR
VELPCM0486000 VELPCM0486001
Kjeldahl weighing boat
Nacelles Nacelles de pesée
Weighing boat is suitable for the syrupy and viscose substances, pulps and powders.
  • Nitrogen-free boat, ensures low error risk
  • Suitable for quantitative transfer of substance into mineralisation test tubes