Imprimer
Chairs
Fournisseur: TOPSTAR
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
72240PU0H 72240PU0T
132-0942EA 0 EUR
132-0942 132-0943
Chairs
Chaises
Chairs with standard gas lift and double-wheel castors/fixed glides.
  • Seat height adjustable in various positions by safety gas lift
  • Backrest adjustable in height and depth with permanent-contact-mechanism
  • Seat and backrest made of durable polyurethane foam, low maintenance
  • Black polyamide base, equipped with braked safety castors for carpet floors
  • With or without foot rings and foot glides
Seat dimension (W×D): 440×430 mm
Backrest height: Backrest: 320 mm