Imprimer
Gel documentation systems, PhotoDoc-It™ 65
Fournisseur: UVP ULTRA VIOLET PRODUCTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
97-0274-05 97-0274-02 97-0274-08 97-0274-11 97-0274-14
732-1249EA 5950 EUR
732-1249 732-1248 732-1226 732-1227 732-1228
Gel documentation systems, PhotoDoc-It™ 65
Photodocumentation Systems
The PhotoDoc-It™ 65 is designed to capture, print and save colour images for basic gel documentation without the need for film.
  • Digi 65 colour camera with mega pixel resolution - view images on the camera or the optional colour LCD screen
  • Lightweight hood provides a compact darkroom environment
  • Save images to memory card or instantly generate 10×15 cm (4×6") low cost colour prints using the photo printer included
  • Ethidium bromide filter for fluorescent stained gels (EtBr Red 570 - 640 nm), positioned inside hood, is easily interchangeable for other filter types
  • Combination lock secures the camera to the darkroom
W×D×H: 337×241×432 mm
Informations de livraison: System supplied with inclusive colour printer and available with or without Benchtop UV transilluminator.