Imprimer
Rotors for Minispin®/Minispin® Plus Centrifuges
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5452720.008 5452727.007
EPPE5452720.008EA 302 EUR
EPPE5452720.008 521-1585
Rotors for Minispin®/Minispin® Plus Centrifuges
Centrifugeuses Rotors
Eppendorf Rotors for MiniSpin/MiniSpin plus, precision for mini centrifugation. Tailor your experiments with varied capacities and speeds. Accessorise with versatile adaptors and buckets for efficient sample processing in a compact format.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
Elevate your mini centrifugation experience with Eppendorf rotors designed for MiniSpin/MiniSpin plus. These rotors offer a range of capacities and speeds, catering to diverse sample processing needs. Accessorise your setup with versatile adaptors and buckets, enhancing the flexibility of your mini centrifuge for various applications. Achieve precision in your experiments within the compact format, ensuring reliable and efficient sample separation. Explore the comprehensive range to optimise your MiniSpin/MiniSpin plus for reproducible results.