Imprimer
Protease inhibitor cocktail, TCM-ProteaseArrest™
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
786-239 786-238
GENO786-239EA 681 EUR
GENO786-239 GENO786-238
Protease inhibitor cocktail, TCM-ProteaseArrest™
Suppléments et additifs pour culture cellulaire
TCM-ProteaseArrest™ is a mixture of protease inhibitors with a broad specificity for the inhibition of serine, cysteine, aspartic (acid) proteases and aminopeptidases.
  • For tissue culture medium to prevent the degradation of secreted proteins
  • Store at –20 °C
TCM-ProteaseArrest™ is supplied as a 200X concentrated solution in sterile filtered DMSO. It is highly recommended to determine the appropriate dilution factor for a specific cell line, starting at a 1:200 dilution of TCM-ProteaseArrest™ to prevent harmful effects of DMSO to cell growth.