Imprimer
Cut resistant gloves, Showa 541
Fournisseur: Showa
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
541/L 541/M 541/S 541/XL 541/XXL
112-5861EA 173 EUR
112-5861 112-5862 112-5863 112-5864 112-5865
Cut resistant gloves, Showa 541
Gants
These seamless knitted HPPE liners with PU coating on palm and fingers provide a high degree of comfort when performing delicate tasks in greasy work environments with a high risk of cuts.
  • HPPE lining provides extremely comfortable fit
  • Knitwrist secures glove above the wrist for comfort and prevents debris from entering the glove
  • Flat-dipped, grey colored PU coating
  • Launderable
Typical applications include: Transport, metallurgy, logistics, steel erecting, construction, automotive.
Certifications: EN 388:2016 (4342B); Cat. III