Imprimer
pH indicator, bromo thymol bluel tablets, Lovibond®
Marques : Lovibond
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
511640BT 511641BT
LOVI511640BTEA 49.2 EUR
LOVI511640BT LOVI511641BT
pH indicator, bromo thymol bluel tablets, Lovibond®
Kits de tests environnementaux
Indicator for pH range 6,0 to 7,6.
Can be used for measuring pH in conjunction with the Comparator 2000+ system.