Imprimer
VWR® Hitachi, Xenon lamp
Fournisseur: COLLECTION PREFIX HPLC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
650-1500
903-0155EA 1010 EUR
903-0155 HITA250-1600
VWR® Hitachi, Xenon lamp
Lights Xenon Lights, Lamps and Bulbs
This xenon lamp is suitable for F-2500, F-2700, F-2710, F-4500, F-7000, F-2000, F-3000, F-3010, F-4000 and F-4010 spectrophotometers.