Imprimer
Cryoboxes, PP, D01/D03-series
Fournisseur: GLW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
D03GS D01 D03
GLWAD03GSEA 24 EUR
GLWAD03GS GLWAD01 GLWAD03
Cryoboxes, PP, D01/D03-series
Boîtes Boîtes pour cryotubes
Cryoboxes, PP, natural or coloured, without divider.
  • Autoclavable up to +121 °C and use in freezer down to –90 °C
  • The included clear lid offers a view of the contents without opening the cryobox
  • Without drainage holes in base for catching drips or store fluids (leakproof only at bottom up to 40 mm)
  • Height extension secured with exclusive clicksystem (not glued)
The boxes comes without compartments, in addition they can be used with GLW dividers for individual configuration.They are suitable to store non standarised tubes, stone samples, small items and more. Inner dimension of the box: 124×124 mm, outer dimension of the box: 130×130 mm.
D01-series
Empty box, without divider. Height variable 70 - 80 mm.
D03-series
Empty box, without divider. Height variable 120 - 130 mm.