Imprimer
Wall bracket for bottles and cylinders
Fournisseur: BODE CHEMIE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
980334 980333
115-4026EA 18 EUR
115-4026 115-4024
Wall bracket for bottles and cylinders
Distributeurs de savon et distributeurs de savon liquide
Wall mounted bottle/cylinder holder made from durable plastic and therefore offer a stable solution for multiple applications. The bracket can be quickly and easily attached, cleaned and disinfected.
  • Colour-transparent design
  • Available for 500 ml and 1000 ml bottles and boxes (tissues)
  • Easy to attach to wall
  • Attachment with power strips possible
  • Not autoclavable
Informations de livraison: Delivery includes mounting hardware, installation and operating instructions. Delivery without bottle, canister or dosing pump.