Imprimer
Accessories for magnetic 3D cell culturing, NanoShuttle™-PL
Fournisseur: Greiner Bio-One
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
655841_KIT 657843_3 781841_KIT 655830_1 655840_KIT 657840_KIT 657841_1 657850_3 657846_6 662840_KIT 781830_1 781840_1 655846_KIT 657830_1 662830_1 781846_KIT
734-3336EA 2150 EUR
734-3336 GREI657843_3 GREI781841_KIT GREI655830_1 734-3335 734-3334 GREI657841_1 GREI657850_3 GREI657846_6 734-3340 GREI781830_1 GREI781840_1 GREI655846_KIT GREI657830_1 GREI662830_1 GREI781846_KIT
Accessories for magnetic 3D cell culturing, NanoShuttle™-PL
Bioréacteurs
NanoShuttle™-PL is used to magnetise cells for 3D cell culturing. The cells can be aggregated with magnetic forces, either by levitation or printing, to form structurally and biologically representative 3D models in vitro.
  • Mimicking native tissue environment
  • Rapid 3D model formation within hours
  • No specialised media
  • Easy to handle without sample loss
  • Allows co-culture
With magnetised spheroids, solution addition and removal is made easy by using magnetic force to hold them in a stationary position during aspiration, thereby limiting spheroid loss. Spheroids can also be picked up and transferred between vessels using magnetic tools such as the MagPen™.
NanoShuttle™-PL consists of gold, iron oxide, and poly-L-lysine. NanoShuttle™-PL magnetises cells by electrostatically attaching to cell membranes during an overnight static incubation. Magnetised cells will appear peppered with dark nanoparticles after incubation. NanoShuttle™-PL will stay attached to the cell membrane for up to 8 days, at which point it is released into the 3D culture.