Imprimer
Drying trays
Fournisseur: JUCHHEIM Laborgerate GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VLSOBAYB807801 VLSOBAYB807821 VLSOBAYB807841 VLSOBAYB807861
JUCHVLSOBAYB807801EA 182 EUR
JUCHVLSOBAYB807801 JUCHVLSOBAYB807821 JUCHVLSOBAYB807841 JUCHVLSOBAYB807861
Drying trays
Plateaux/Récipients/Pots Plateaux de séchage
Made of acid resistant stainless steel.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning