Imprimer
Urine collection tubes, primary, with screw cap, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
327220 327230
LSUK327220EA 0 EUR
LSUK327220 LSUK327230
Urine collection tubes, primary, with screw cap, WHEATON®
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons d'urine
These WHEATON® round bottom tubes are manufactured from virgin PS. The tubes have a capacity of 11 ml and are fitted with a leakproof screw cap for ease of transportation and storage of the sample. The tubes are supplied with a collection cup for the patient’s comfort during sample collection, and for them to easily transfer the sample to the tube.
  • Ideal for use with automated urine analysis machines
  • Printed label for patient details
  • Option of plain tube or pre-dosed with boric acid
  • Removes the need for unnecessary decanting of sample in the laboratory
Informations de livraison: Tubes are supplied with sample collection cup.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904