Imprimer
Racks for upright freezers with drawers, extra-wide
Fournisseur: GLW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
W542 W545
GLWAW542EA 342 EUR
GLWAW542 GLWAW545
Racks for upright freezers with drawers, extra-wide
Portoirs Portoirs pour cryoboîtes
These freezer racks are designed for extra wide cryo boxes up to 136×136×54 mm exterior dimension. Suitable for all common upright freezers with various exterior heights.
  • Cryo boxes made of water-repellent fiberboard, cardboard or plastic
  • Racks are made up of stainless steel
  • Each drawer is easily removable and has an ergonomic handle
  • Can be used with the box dimensions 133×133 or 136×136 mm (W×H)