Imprimer
Horizontal racks for chest freezers
Fournisseur: GLW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M133 MD12
479-1605EA 479 EUR
479-1605 479-1606
Horizontal racks for chest freezers
Portoirs Portoirs pour microplaques
These racks are commonly used for chest freezers.
  • Made up of stainless steel
  • For use with deepwell plates and microplates
  • With one folding handle on top
Informations de commande: Optional stainless steel locking rod available separately.