Imprimer
ASTM® precision thermometers, low hazard
Fournisseur: Amarell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A300770-CB/CC4 A318188CC5 A318192CC5 A318132CC3 A318196CC5 A318200CC5 A318204CC5
AMARA300770-CB/CC4EA 0 EUR
AMARA300770-CB/CC4 AMARA318188CC5 AMARA318192CC5 AMARA318132CC3 AMARA318196CC5 AMARA318200CC5 AMARA318204CC5
ASTM® precision thermometers, low hazard
Thermomètres Thermomètres analogiques
Low hazard precision thermometers are used for special applications and suitable for government calibration.
  • Solid stem and white backed
  • Round or prismatic with round top finish
  • Durable pigment
  • Non-wetting because of capillary tube specially coated inside
Certifications: ASTM standard E2251–07 (Standard specification for liquid-in-glass ASTM thermometers with low-hazard precision liquids).
Cdt: Packaged in transparent square plastic case.