Imprimer
Venting needles for high precision sample handling
Fournisseur: HAMILTON BONADUZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
210540 210541 210542 210543 210544 210545 210546 210547 210548 210549 210550 210551 210552 210553 210554 210555
HAMI210540EA 559 EUR
HAMI210540 HAMI210541 HAMI210542 HAMI210543 HAMI210544 HAMI210545 HAMI210546 HAMI210547 HAMI210548 HAMI210549 HAMI210550 HAMI210551 HAMI210552 HAMI210553 HAMI210554 HAMI210555
Venting needles for high precision sample handling
Aiguilles pour seringues
These venting needles are made of high grade stainless steel and are ideally suited for a wide range of applications. Various lengths and different hub connections are available to perfectly match the requirements of laboratory equipment manufacturers.
  • OEM customisation
  • More accurate pipetting with high corrosion resistance
  • Prevents cross-contamination
  • Self-venting design allows pressure compensation
  • Aspirate more than 50 μl in one injection
  • Process more samples in shorter time
These needles are designed to address the over and under-pressurisation that arises during precise dispensing and aspirating in a closed vessel.
Point style 5: Conical with side port for penetration without coring. Applications prone to needle clogging; causes minimal septum damage.