Imprimer
Chemkey® Reagents
Marques : Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9425200 9429100 9429300 9429000 9429200 9429400 9429600 8499300 8499400 8636000 8636100 8636200 8636300 8636400 8636500 8636600 8791900 9429500
705-0952EA 93.2 EUR
705-0952 705-0945 705-0949 705-0943 705-0953 705-0947 705-0948 705-0944 705-0946 705-0954 705-0955 705-0956 705-0957 705-0958 705-0959 705-0960 HACH8791900 HACH9429500
Chemkey® Reagents
Kits de tests environnementaux
Chemkey® reagents are a new technology that enable the SL1000 Portable Parallel Analyzer colorimeter to perform the same tests with less than half the manual steps.
  • All chemicals and processes are entirely contained inside the Chemkey®
  • There are no powder pillows or glass vials to handle
  • Get highly accurate results with less opportunity for errors in a fraction of the time
There is no zeroing, no mixing, no shaking, no chemicals or vials to handle. All you have to do is insert the chemkey and let the PPA-SL1000 read you back the results.