Imprimer
Standards for water pollution QC check, solids, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-QWPSOL-23 VHG-QWSSOL-23
PROOVHG-QWPSOL-23EA 457 EUR
PROOVHG-QWPSOL-23 PROOVHG-QWSSOL-23
Standards for water pollution QC check, solids, VHG
Étalons Étalons pour l'analyse environnementale
This product is intended for use as a quality control check for methods related to total suspended solids (TSS), total dissolved solids (TDS), and total solids (TS).
This standard has been prepared as a concentrate and must be diluted prior to analysis. This standard is not preserved but can be stored at room temperature.
Certifications: Traceable to NIST SRM.