Imprimer
Tungsten standard solution, for ICP, in H₂O, CRM, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-PWW-100 VHG-PWW-500 VHG-TWW-500 VHG-TWW-100
PROOVHG-PWW-100EA 275 EUR
PROOVHG-PWW-100 PROOVHG-PWW-500 PROOVHG-TWW-500 PROOVHG-TWW-100
Tungsten standard solution, for ICP, in H₂O, CRM, VHG
Étalons Étalons cationiques Standards de tungstène
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.