Imprimer
Uranium standard solution, for ICP, in HNO₃, CRM, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-PUN-100 VHG-PUN-500 VHG-PUN-50
PROOVHG-PUN-100EA 265 EUR
PROOVHG-PUN-100 PROOVHG-PUN-500 PROOVHG-PUN-50
Uranium standard solution, for ICP, in HNO₃, CRM, VHG
Étalons Étalons cationiques Standards d'uranium
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.