Imprimer
Tissue grinders, CS1, KIMBLE®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SP749600-0015 SP749600-0050
SCERSP749600-0015EA 313 EUR
SCERSP749600-0015 SCERSP749600-0050
Tissue grinders, CS1, KIMBLE®
Homogénéisateurs Broyeurs pour tissus
CS1 tissue grinders are suitable for standard grinding, tissues are ground within a sealed container to minimise the risk of personal contact.
  • Narrow pestles eliminate the potential of sample overflow
  • Convenient and easy-to-use
  • Gamma sterilised in an easy-open Tyvek® pack
The molded-in abrasive surface on the pestle tip leaves no sediment to obstruct sample examination. Tubes are made of PP, pestles are 30% glass-filled PP and caps are HDPE.
Informations de livraison: Each tissue grinder is supplied with a fully assembled pestle, a conical sample tube with solid top cap, an adhesive identification label and an instruction card.